Learn More
D-Ribulose (~1M aqueous solution), TRC
Specifications
Specifications
| Chemical Name or Material | D-Ribulose |
| CAS | 488-84-6 |
| Molecular Formula | C5 H10 O5 |
| Quantity | 50 mg |
| Synonym | D-erythro-2-Pentulose, D-erythro-Pentulose (7CI,8CI), D(-)-Ribulose, D-Adonose, D-Arabinulose, D-Araboketose, D-Erythropentulose, D-Ribosone, D-Ribulose, D-erythro-2-Ketopentose |
| InChI Formula | InChI=1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h3,5-8,10H,1-2H2/t3-,5-/m1/s1 |
| SMILES | OC[C@@H](O)[C@@H](O)C(=O)CO |
| IUPAC Name | (3R,4R)-1,3,4,5-tetrahydroxypentan-2-one |
| Molecular Weight (g/mol) | 150.13 |
| Formula Weight | 150.0528 |