Learn More
D-Phenylalaninol, 98%, Thermo Scientific Chemicals
Product Code. p-7045177
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11352217 250 mg 250mg
11342217 1 g 1g
2 options
Product Code. 11352217
Brand: Thermo Scientific Alfa Aesar L09697.MD
Request Bulk or Custom Format
D-Phenylalaninol is used as a chiral auxiliary for asymmetric Michael reactions. It is an enantiomer of L-Phenylalaninol, an inhibitor of intestinal Phenylalanine absorption. It acts as an inhibiting agent to the enzymes which are responsible for the breakdown of endorphins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Phenylalaninol is used as a chiral auxiliary for asymmetric Michael reactions. It is an enantiomer of L-Phenylalaninol, an inhibitor of intestinal Phenylalanine absorption. It acts as an inhibiting agent to the enzymes which are responsible for the breakdown of endorphins.
Solubility
Soluble in Dichloromethane, Ethyl Acetate, Methanol.
Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with carbondioxide, air, acids and oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5267-64-1
Molecular Formula C9H13NO
MDL Number MFCD00064298
Synonym d-phenylalaninol, r-2-amino-3-phenylpropan-1-ol, d-penylalaninol, r-+-2-amino-3-phenyl-1-propanol, 2r-2-amino-3-phenylpropan-1-ol, d +-phenylalaninol, +-d-phenylalaninol, r-2-amino-3-phenyl-1-propanol, d-+-phenylalaninol, r-phenylalaninol
Specifications
Melting Point 89°C to 94°C
Quantity 250 mg
Optical Rotation +23 ± 2° (c=5 in Ethanol)
UN Number UN3259
Beilstein 4665408
Formula Weight 151.21
Percent Purity 98%
Chemical Name or Material D-Phenylalaninol