missing translation for 'onlineSavingsMsg'
Learn More

D-myo-Inositol, 99.2%, Cell culture reagent, For HPLC analysis, MP Biomedicals™

Product Code. 11461881 Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 g
100 g
1 kg
Unit Size:
100g
1kg
50g
This item is not returnable. View return policy

Product Code. 11461881

Brand: MP Biomedicals 0219468880

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00077932
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
Synonym scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
PubChem CID 892
ChEBI CHEBI:24848
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES C1(C(C(C(C(C1O)O)O)O)O)O

Specifications

Color White
Density 6.2g/mL
Quantity 100 g
Formula Weight 180.2
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material D-myo-inositol, Molecular biology and Cell biology
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.