missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Maltose monohydrate, 95 to 98%, MP Biomedicals™

Catalog Number p-4679772
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy
6363-53-7
C12H24O12
360.31
MFCD00149343
HBDJFVFTHLOSDW-UHFFFAOYNA-N
d-+-maltose monohydrate, unii-dm477ee40d, 2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate, beta-maltose monohydrate, d-+-maltosemonohydrate, 69-79-4 anhydrous, d +-maltose monohydrate, d-+-maltose monohydrate, puriss., d-+-maltose monohydrate, analytical standard, d-+-maltose monohydrate, bioxtra
2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O
This item is not returnable. View return policy
6363-53-7
C12H24O12
360.31
MFCD00149343
HBDJFVFTHLOSDW-UHFFFAOYNA-N
d-+-maltose monohydrate, unii-dm477ee40d, 2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate, beta-maltose monohydrate, d-+-maltosemonohydrate, 69-79-4 anhydrous, d +-maltose monohydrate, d-+-maltose monohydrate, puriss., d-+-maltose monohydrate, analytical standard, d-+-maltose monohydrate, bioxtra
2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O
Molecular biology
Color White
Density 1.54g/mL
Assay Percent Range 95 to 98%
Quantity 100 g
Formula Weight 360.32
Physical Form Solid
Chemical Name or Material D-(+)-Maltose monohydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'