Learn More
D-Leucine, 99%, Thermo Scientific Chemicals
Product Code. p-7022195
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11402617
Brand: Thermo Scientific Alfa Aesar A14842.06
Request Bulk or Custom Format
D-Leucine is used in the formation of sterols. It is also used as a selectable marker.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Leucine is used in the formation of sterols. It is also used as a selectable marker.
Solubility
Soluble in water and hydrochloric acid.
Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 328-38-1
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00063088
InChI Key ROHFNLRQFUQHCH-RXMQYKEDSA-N
Synonym d-leucine, h-d-leu-oh, r-leucine, 2r-2-amino-4-methylpentanoic acid, d-homo-valine, r---leucine, leucine, d, r-2-amino-4-methylpentanoic acid, d-leucin, d-leuzin
PubChem CID 439524
ChEBI CHEBI:28225
IUPAC Name (2R)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@@H](N)C(O)=O
Specifications
Melting Point >300°C
Beilstein 1721721
Solubility Information Soluble in water and hydrochloric acid.
Quantity 5 g
Formula Weight 131.18
Optical Rotation −15° (c=5 in 5N HCl)
Percent Purity 99%
Chemical Name or Material D-Leucine