Learn More
D,L-Venlafaxine-d6, TRC
Specifications
Specifications
| Chemical Name or Material | D,L-Venlafaxine-d6 |
| CAS | 1020720-02-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17H21D6NO2 |
| Quantity | 50 mg |
| Synonym | 1-[2-(Dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexanol-d6 |
| InChI Formula | InChI=1S/C17H27NO2/c1-18(2)13-16(17(19)11-5-4-6-12-17)14-7-9-15(20-3)10-8-14/h7-10,16,19H,4-6,11-13H2,1-3H3/i4D2,5D2,6D2 |
| SMILES | OC1(CC([2H])([2H])C([2H])([2H])C([2H])([2H])C1)C(CN(C)C)C2=CC=C(OC)C=C2 |
| IUPAC Name | 3,3,4,4,5,5-hexadeuterio-1-[2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol |
| Molecular Weight (g/mol) | 283.44 |
| Show More |