Learn More
D(+)-Glucuronic Acid Sodium Salt Monohydrate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 207300-70-7 |
| Melting Point | 137°C to 141°C |
| MDL Number | MFCD12966022 |
| Quantity | 10 g |
| Synonym | D(+)-Glucuronic acid sodium salt monohydrate, Sodium D-glucuronate, D(+)-Glucuronic acid sodium salt monohydrate |
| Molecular Weight (g/mol) | 216.12 (Anhydrous Basis) |
| Grade | Research |