Learn More
D-Glucose dehydrogenase, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 kU
Unit Size:
5kg
Description
Diagnostics and cofactor regeneration of NAD(P)H in reductive processes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS | 9028-53-9 |
| Color | Yellow |
| MDL Number | MFCD00131181 |
| Quantity | 5 kU |
| Physical Form | Powder |
| Chemical Name or Material | D-Glucose dehydrogenase |