missing translation for 'onlineSavingsMsg'
Learn More

D-Glucosamine sulfate, MP Biomedicals™

Product Code. 11477362 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477362 25 g 25g
11467362 10 g 10g
11487362 100 g 100g
11497362 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11477362 Supplier MP Biomedicals Supplier No. 0215392825

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 29031-19-4
Molecular Formula C6H15NO9S
Molecular Weight (g/mol) 277.244
InChI Key FGNPLIQZJCYWLE-BTVCFUMJSA-N
Synonym d-glucosamine sulphate, 2r,3r,4s,5r-2-amino-3,4,5,6-tetrahydroxyhexanal sulfate, gevolox, glucosaminsulfat, sulfato de glucosamina, 2-amino-2-deoxy-d-glucose sulfate salt, glucosamine sulfate 2kcl, d-glucose, 2-amino-2-deoxy-, sulfate salt, glucosamine sulfate na salt, glucosamine sulfate dc
PubChem CID 115046
IUPAC Name (2R,3R,4S,5R)-2-amino-3,4,5,6-tetrahydroxyhexanal;sulfuric acid
SMILES C(C(C(C(C(C=O)N)O)O)O)O.OS(=O)(=O)O

Specifications

Quantity 25 g
Formula Weight 228.2
Physical Form Solid
Chemical Name or Material D-Glucosamine Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.