Learn More
D(-)-Fructose, Millipore™
Product Code. 10762442
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10762442 1 kg 1kg
10578470 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10762442 Supplier Millipore Supplier No. 1040071000
 Restricted Item
Estimated Shipment: 26-11-2026
For biochemistry
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 57-48-7
Specifications
Melting Point 100°C to 104°C (decomposition)
Density 1.59 g/cm3 at 20°C (OECD Test Guideline 109)
MDL Number MFCD00148910
Quantity 1 kg
Synonym D(-)-Fructose, Laevulose, Levulose, D(-)-Fructose
Molecular Weight (g/mol) 180.16
Grade Research