missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Dimer Antibody (DD2) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB110-8376JF525
This item is not returnable.
View return policy
Description
D-Dimer Monoclonal antibody specifically detects D-Dimer in Human samples. It is validated for ELISA,Western Blot,Immunoassay,Sandwich ELISA
Specifications
| D-Dimer | |
| Monoclonal | |
| Janelia Fluor 525 | |
| D Dimer | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with D-dimer. | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b |
| ELISA, Western Blot, Immunoassay, Sandwich ELISA | |
| DD2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction