Learn More
IBA Lifesciences Desthiobiotin
GREENER_CHOICE
Product Code. p-8014578 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
Each
Product Code. Quantity unitSize
12753064 1 g Each
12763064 5 g Each
2 options
Product Code. 12753064
Brand: IBA Lifesciences 21000002
Edge
Desthiobiotin powder for preparation of Strep-Tactin™ elution buffer
Desthiobiotin is a modified form of biotin that displaces Strep-tag™II and Twin-Strep-tag™ fusion proteins from the modified biotin-binding pocket of Strep-Tactin™ in a competitive manner, resulting in a mild elution.
Desthiobiotin powder can be used for preparation of Strep-Tactin™ elution buffer, Buffer E.
Please note that desthiobiotin is not applicable for elution from Strep-Tactin™XT resins.
Specifications
Chemical Name or Material Desthiobiotin
For Use With (Application) For Elution of Strep-Tag II Proteins From Strep-Tactin
Quantity 1 g
For Research Use Only.