Learn More
D-Cysteine, TRC
Product Code. 30179378
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179378 25 g 25g
30179668 1 g 1g
30179394 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30179378 Supplier Toronto Research Chemicals Supplier No. TRCC99502025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 921-01-7
Molecular Formula C3 H7 N O2 S
Molecular Weight (g/mol) 121.16
Synonym D-Cysteine (9CI, ACI), Cysteine, D- (8CI), (2S)-2-Amino-3-sulfanylpropanoic acid, (S)-2-Amino-3-mercaptopropanoic acid, (S)-Cysteine
IUPAC Name (2S)-2-amino-3-sulfanylpropanoic acid
SMILES N[C@H](CS)C(=O)O
Specifications
InChI Formula InChI=1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)/t2-/m1/s1
Quantity 25 g
Formula Weight 121.0197
Shipping Condition Room Temperature
Chemical Name or Material D-Cysteine