Learn More
D-Camphor, Mikromol™
Product Code. 17981426
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17981426 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17981426 Supplier Mikromol Supplier No. MM3327.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
D-Camphor
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (+)-Camphor
CAS 464-49-3
Analyte or Component Names (+)-Camphor
Shipping Condition Room Temperature
Molecular Formula C10 H16 O
Certifications/Compliance ISO 9001
Synonym d-Camphor, (1R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one
InChI Formula InChI=1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m1/s1
SMILES CC1(C)[C@@H]2CC[C@@]1(C)C(=O)C2
IUPAC Name (1R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
Molecular Weight (g/mol) 152.23
Quantity 250 mg
Formula Weight 152.1201 g/mol