Learn More
D-Camphor, LoGiCal
Product Code. 30459411
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459411 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30459411 Supplier LoGiCal Supplier No. LGCFOR3327.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (+)-Camphor
CAS 464-49-3
Analyte or Component Names (+)-Camphor
Shipping Condition Room Temperature
Molecular Formula C10 H16 O
Certifications/Compliance ISO 9001
Synonym d-Camphor, (1R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one
InChI Formula InChI=1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m1/s1
Solution Type Neat
SMILES CC1(C)[C@@H]2CC[C@@]1(C)C(=O)C2
IUPAC Name (1R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
Industry Type Clinical Testing
Molecular Weight (g/mol) 152.23
Quantity 10 mg
Formula Weight 152.1201 g/mol