missing translation for 'onlineSavingsMsg'
Learn More

D-beta-Proline ethyl ester hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number 15464208
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1807350-90-8
C7H14ClNO2
179.644
MFCD18071037
AYYKCALHTPKNOI-FYZOBXCZSA-N
ethyl r-3-pyrrolidinecarboxylate hydrochloride, d-beta-proline ethyl ester hydrochloride, r-ethyl pyrrolidine-3-carboxylate hydrochloride, ethyl r-3-pyrrolidinecarboxylate, hcl, r-ethyl pyrrolidine-3-carboxylate hcl, ethyl r---pyrrolidine-3-carboxylate hydrochloride, ethyl r-pyrrolidine-3-carboxylate hydrochloride, ethyl 3r-pyrrolidine-3-carboxylate hydrochloride, r---pyrrolidine-3-carboxylic acid ethyl ester hydrochloride
53488453
ethyl (3R)-pyrrolidine-3-carboxylate;hydrochloride
CCOC(=O)C1CCNC1.Cl
This item is not returnable. View return policy
1807350-90-8
C7H14ClNO2
179.644
MFCD18071037
AYYKCALHTPKNOI-FYZOBXCZSA-N
ethyl r-3-pyrrolidinecarboxylate hydrochloride, d-beta-proline ethyl ester hydrochloride, r-ethyl pyrrolidine-3-carboxylate hydrochloride, ethyl r-3-pyrrolidinecarboxylate, hcl, r-ethyl pyrrolidine-3-carboxylate hcl, ethyl r---pyrrolidine-3-carboxylate hydrochloride, ethyl r-pyrrolidine-3-carboxylate hydrochloride, ethyl 3r-pyrrolidine-3-carboxylate hydrochloride, r---pyrrolidine-3-carboxylic acid ethyl ester hydrochloride
53488453
ethyl (3R)-pyrrolidine-3-carboxylate;hydrochloride
CCOC(=O)C1CCNC1.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Protect from heat. Store away from oxidizing agents. Refrigerate. Incompatible with heat and oxidizing agents.
TRUSTED_SUSTAINABILITY
Formula Weight 179.65
Percent Purity 97%
Quantity 250 mg
Optical Rotation −14° (c=0.6 in Water)
Chemical Name or Material D-beta-Proline ethyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'