Learn More
D-Alanine-2-d1, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | D-Alanine-2-D |
| CAS | 56595-54-1 |
| Molecular Formula | C3 D H6 N O2 |
| Quantity | 0.25 g |
| Synonym | D-Alanine-2-d (9 CI, ACI), (2 R)-2-Amino-2-deuteriopropanoic acid, D-(α2H)Alanine, (2 R)-2-Aminopropanoic-2-d acid, D-Alanine-2-D, D-[2-2H1]Alanine |
| InChI Formula | InChI=1 S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m1/s1/i2D |
| SMILES | [2 H][C@](C)(N)C(=O)O |
| IUPAC Name | (2 R)-2-amino-2-deuteriopropanoic acid |
| Molecular Weight (g/mol) | 90.0993 |
| Formula Weight | 90.054 g/mol |
| Show More |