Learn More
D-Abequose, TRC
Product Code. 30179589
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179589 2.5 mg 2.5mg
30179337 10 mg 10mg
30179133 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30179589 Supplier Toronto Research Chemicals Supplier No. TRCA0102052.5MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 56816-60-5
Molecular Formula C6 H12 O4
Molecular Weight (g/mol) 148.16
Synonym Abequose (6CI), 3,6-Dideoxy-D-xylo-hexose;
IUPAC Name (2R,4R,5R)-2,4,5-trihydroxyhexanal
SMILES C[C@@H](O)[C@H](O)C[C@@H](O)C=O
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C6H12O4/c1-4(8)6(10)2-5(9)3-7/h3-6,8-10H,2H2,1H3/t4-,5-,6-/m1/s1
Formula Weight 148.0736
Chemical Name or Material D-Abequose