Learn More
Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips)
GREENER_CHOICE
Product Code. 12917207
Change view
Click to view available options
Volume (Metric):
12.5 μL
30 μL
100 μL
220 μL
250 μL
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Packaging:
10 x 384 tips
20 x 384 tips
20 x 96 tips
Unit Size:
Pack of 1920
Pack of 3840
Pack of 7680
Product Code. Volume (Metric) Sterility Filtered Packaging unitSize
12917207 30 μL Sterile Non-filtered 20 x 96 tips Pack of 1920
10089982 12.5 μL Non-sterile Non-filtered 10 x 384 tips Pack of 3840
10794434 12.5 μL Sterile Non-filtered 10 x 384 tips Pack of 3840
10592285 30 μL Sterile Non-filtered 20 x 384 tips Pack of 7680
10502295 30 μL Sterile Filtered 20 x 384 tips Pack of 7680
10283293 100 μL Non-sterile Non-filtered 10 x 384 tips Pack of 3840
12987197 100 μL Non-sterile Non-filtered 20 x 384 tips Pack of 7680
11719048 100 μL Sterile Non-filtered 20 x 384 tips Pack of 7680
10203443 100 μL Sterile Filtered 20 x 384 tips Pack of 7680
10573625 100 μL Non-sterile Non-filtered 20 x 96 tips Pack of 1920
11567459 100 μL Sterile Non-filtered 20 x 96 tips Pack of 1920
11577459 100 μL Sterile Filtered 20 x 96 tips Pack of 1920
11507479 30 μL Non-sterile Non-filtered 20 x 96 tips Pack of 1920
10572285 30 μL Sterile Non-filtered 20 x 96 tips Pack of 1920
10532865 30 μL Sterile Filtered 20 x 96 tips Pack of 1920
10541715 30 μL Non-sterile Non-filtered 10 x 384 tips Pack of 3840
10485953 220 μL Non-sterile Non-filtered 20 x 96 tips Pack of 1920
10495953 250 μL Sterile Non-filtered 20 x 96 tips Pack of 1920
10101633 220 μL Sterile Filtered 20 x 96 tips Pack of 1920
10747314 30 μL Non-sterile Non-filtered 20 x 96 tips Pack of 1920
10171393 100 μL Sterile - 10 x 384 tips Pack of 3840
10355073 30 μL Non-sterile Non-filtered 20 x 384 tips Pack of 7680
22 options
Product Code. 12917207
Brand: Thermo Scientific™ 5572
Only null left
Add to basket
Add to basket
Achieve ultimate performance in automated pipetting with these tips--superior quality with the flexibility to choose tips for any application.
Perform pipetting more effectively with Thermo Scientific™ Matrix™ SerialMate™ Pipette Tips, for use with the Matrix SerialMate™ Pipetter. Unlike conventional automation tips, D.A.R.T.'s™ are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision.
Features:
  • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access.
  • Face seal design ensures consistent tip-to-tip height for precise low volume dispensing
  • Highly polished molds and cores ensure low liquid retention.
  • Small internal tip orifice provides better small volume droplet formation, resulting in optimal accuracy and precision.
  • Sterile, sterile filtered, wide-bore, and extended-length tip options are available
  • Tips are thoroughly inspected for proper formation and straightness
  • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing.
  • Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination.
Includes: 20 magazines per case
Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x2, Matrix™ PlateMate™ 2x3, PlateMate™ Plus Systems and Versette.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 30 μL
Filtered Non-filtered
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001 and ISO 13485
Clean Claims Free of RNase, DNase, DNA, ATP and endotoxin
Format Rack
Tip Style Automation
Packaging 20 x 96 tips
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Disposable Single-use
For Use With (Equipment) Thermo Scientific Matrix PlateMate, Thermo Scientific Matrix Hydra DT
Product Line D.A.R.T.S