Learn More
Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips)
GREENER_CHOICE
Product Code. 10089982
Change view
Click to view available options
Volume (Metric):
12.5 μL
30 μL
100 μL
220 μL
250 μL
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Packaging:
10 x 384 tips
20 x 384 tips
20 x 96 tips
Unit Size:
Pack of 1920
Pack of 3840
Pack of 7680
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Filtered Packaging unitSize
10089982 12.5 μL Non-sterile Non-filtered 10 x 384 tips Pack of 3840
10794434 12.5 μL Sterile Non-filtered 10 x 384 tips Pack of 3840
10592285 30 μL Sterile Non-filtered 20 x 384 tips Pack of 7680
10502295 30 μL Sterile Filtered 20 x 384 tips Pack of 7680
10283293 100 μL Non-sterile Non-filtered 10 x 384 tips Pack of 3840
12987197 100 μL Non-sterile Non-filtered 20 x 384 tips Pack of 7680
11719048 100 μL Sterile Non-filtered 20 x 384 tips Pack of 7680
10203443 100 μL Sterile Filtered 20 x 384 tips Pack of 7680
10573625 100 μL Non-sterile Non-filtered 20 x 96 tips Pack of 1920
11567459 100 μL Sterile Non-filtered 20 x 96 tips Pack of 1920
11577459 100 μL Sterile Filtered 20 x 96 tips Pack of 1920
11507479 30 μL Non-sterile Non-filtered 20 x 96 tips Pack of 1920
10572285 30 μL Sterile Non-filtered 20 x 96 tips Pack of 1920
10532865 30 μL Sterile Filtered 20 x 96 tips Pack of 1920
10541715 30 μL Non-sterile Non-filtered 10 x 384 tips Pack of 3840
10485953 220 μL Non-sterile Non-filtered 20 x 96 tips Pack of 1920
10495953 250 μL Sterile Non-filtered 20 x 96 tips Pack of 1920
10101633 220 μL Sterile Filtered 20 x 96 tips Pack of 1920
10747314 30 μL Non-sterile Non-filtered 20 x 96 tips Pack of 1920
10171393 100 μL Sterile - 10 x 384 tips Pack of 3840
12917207 30 μL Sterile Non-filtered 20 x 96 tips Pack of 1920
10355073 30 μL Non-sterile Non-filtered 20 x 384 tips Pack of 7680
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10089982 Lieferant Thermo Scientific™ Lieferanten-Nr. 5301
  • ONLINE EXCLUSIVE PRICE
    11090.00 NOK
    10809.67 NOK / Pack of 3840
    Save 280.33 NOK
    2% Off
Alle Werbeaktionen anzeigen
Estimated Shipment: 23-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Achieve ultimate performance in automated pipetting with these tips--superior quality with the flexibility to choose tips for any application.
Perform pipetting more effectively with Thermo Scientific™ Matrix™ SerialMate™ Pipette Tips, for use with the Matrix SerialMate™ Pipetter. Unlike conventional automation tips, D.A.R.T.'s™ are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision.
Features:
  • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access
  • Highly polished molds and cores ensure low liquid retention
  • Small internal tip orifice provides better small volume droplet formation
  • Sterile, sterile-filtered, wide-bore, and extended-length tip options are available
  • Tips are thoroughly inspected for proper formation and straightness
  • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing
  • Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination
Includes: 20 magazines per case
Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x3 and PlateMate™ Plus Systems.
TRUSTED_SUSTAINABILITY
Spezifikation
Description Standard length
Volume (Metric) 12.5 μL
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Certifications/Compliance ISO 9001 and ISO 13485
Clean Claims Free of RNase, DNase, DNA, ATP, and endotoxin
Format Rack
Tip Style Automation
Packaging 10 x 384 tips
Quantity 3840 tips
Disposable Single-use
For Use With (Equipment) Thermo Scientific Matrix Hydra DT, Thermo Scientific PlateMate Plus
Product Line D.A.R.T.S, Matrix
Mehr anzeigen Weniger anzeigen