missing translation for 'onlineSavingsMsg'
Learn More

Cytosine-β-D-Arabinofuranoside, Hydrochloride, MP Biomedicals™

Product Code. 11328369 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328369 5 g 5g
11318369 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328369 Supplier MP Biomedicals Supplier No. 0210007105
  • ONLINE EXCLUSIVE PRICE
    14275.00 NOK
    13598.89 NOK / 5g
    Save 676.11 NOK
    4% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An anti-leukemic agent resulting in DNA fragmentation and inhibition of replication

Identificatori chimici

CAS 69-74-9
Molecular Formula C9H14ClN3O5
Molecular Weight (g/mol) 279.68
MDL Number MFCD00012839
InChI Key KCURWTAZOZXKSJ-JBMRGDGGSA-N
Synonym cytarabine hydrochloride
PubChem CID 134129637
IUPAC Name 4-amino-1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one hydrochloride
SMILES Cl.NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O

Specifica

Color White
Quantity 5 g
Formula Weight 279.7
Physical Form Powder
Chemical Name or Material Cytosine-β-D-Arabinofuranoside, Hydrochloride
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.