Learn More
CytoScint ES, MP Biomedicals™
Product Code. 15702252 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 gal.
4 x 1 gal.
Unit Size:
15.14 L
3.79 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15702252 1 gal. 3.79 L
15712252 4 x 1 gal. 15.14 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15702252 Supplier MP Biomedicals Supplier No. 0188245301
  • ONLINE EXCLUSIVE PRICE
    4200.00 NOK
    4000.41 NOK / 3.79 L
    Save 199.59 NOK
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket
Liquid Scintillation Cocktails; CytoScint ES; MP Biomedical; 4 x 1 gal.; Biodegradable
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Liquid Scintillation Cocktails
Quantity 1 gal.
compliance-icons compliance-icons
Product Identifier
  • cytoscint ES
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: phenylxylylethane, linear alkylbenzene,ethoxylated nonylphenol, 2-methoxyethanol