missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 8 Antibody (KRT8/803), Novus Biologicals™
Mouse Monoclonal Antibody
2535.75 NOK - 4332.30 NOK
Specifications
| Antigen | Cytokeratin 8 |
|---|---|
| Clone | KRT8/803 |
| Concentration | 0.2mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18139587
|
Novus Biologicals
NBP2-44940-0.02MG |
0.02 mg |
2685.00 NOK 2535.75 NOK / 0.02mg Save 149.25 NOK 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18160898
|
Novus Biologicals
NBP2-44940-0.1MG |
0.1 mg |
4580.00 NOK 4332.30 NOK / 0.10mg Save 247.70 NOK 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Cytokeratin 8 Monoclonal specifically detects Cytokeratin 8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Cytokeratin 8 | |
| 0.2mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Cytoskeleton Markers | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 3856 | |
| Recombinant human KRT8 protein | |
| Primary | |
| Store at 4C. | |
| 52.5 kDa |
| KRT8/803 | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
| KRT8 | |
| IgG1 κ | |
| Protein A or G purified | |
| Cytokeratin 8 (CK8) belongs to the type II (or B or basic) subfamily of high molecular weight cytokeratins and exists in combination with cytokeratin 18 (CK18). CK8 is primarily found in the non-squamous epithelia and is present in majority of adenocarcinomas and ductal carcinomas. It is absent in squamous cell carcinomas. Hepatocellular carcinomas are defined by the use of antibodies that recognize only cytokeratin 8 and 18. CK8 exists on several types of normal and neoplastic epithelia, including many ductal and glandular epithelia such as colon, stomach, small intestine, trachea, and esophagus as well as in transitional epithelium. Anti-CK8 does not react with skeletal muscle or nerve cells. Epithelioid sarcoma, chordoma, and adamantinoma show strong positivity corresponding to that of simple epithelia (with antibodies against CK8, CK18 and CK19). Anti-CK8 is useful for the differentiation of lobular (ring-like, perinuclear) from ductal (peripheral-predominant) carcinoma of the breast. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title