Learn More
Cytokeratin 7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15539-1-AP-20UL
This item is not returnable.
View return policy
Description
Cytokeratin 7 blocks interferon-dependent interphase and stimulates DNA synthesis in cells. Involved in the translational regulation of the human papillomavirus type 16 E7 mRNA (HPV16 E7).Specifications
| Cytokeratin 7 | |
| Polyclonal | |
| Unconjugated | |
| KRT7 | |
| CK 7, CK7, Cytokeratin 7, K2C7, K7, keratin 7, KRT7, Sarcolectin, SCL, Type II keratin Kb7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 110310, 300242, 3855 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08729, Q6IG12, Q9DCV7 | |
| KRT7 | |
| Cytokeratin 7 Fusion Protein Ag7895 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |