Learn More
Cytokeratin 5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Product Code. 16836175 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16836175 150 μL 150µL
16846175 20 μL 20µL
2 options
Product Code. 16836175
Brand: Proteintech 285061AP150UL
Rabbit Polyclonal Antibody
The Cytokeratin 5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Cytokeratin 5. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 5 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
Antigen Cytokeratin 5
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA
Classification Polyclonal
Concentration 767 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 3852
Target Species Human, Rat, Mouse
Isotype IgG