missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 20 Mouse anti-Human, Clone: 4D10A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60183-1-IG-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. This cytokeratin is a major cellular protein of mature enterocytes and goblet cells and is specifically expressed in the gastric and intestinal mucosa. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.Specifications
| Cytokeratin 20 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35900 | |
| KRT20 | |
| Cytokeratin 20 Fusion Protein Ag11336 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 4D10A4 | |
| Unconjugated | |
| KRT20 | |
| CK 20, CK20, Cytokeratin 20, K20, keratin 20, KRT20, KRT21, Protein IT | |
| Mouse | |
| Protein A | |
| RUO | |
| 54474 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction