missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 19 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10712-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16847273

  • 3828.53 NOK / 150µL
Estimated Shipment: 13-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cytokeratin 19 is part of a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight from 40-68 kDa and isoelectric pH between 4. 9-7. 8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial maligcies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. For example, cytokeratin 19 is not expressed in hepatocytes, therefore, it is useful in the identification of liver metastasis.
Specifications

Specifications

Cytokeratin 19
Polyclonal
Unconjugated
KRT19
CK 19, CK19, Cytokeratin 19, K19, K1CS, keratin 19, KRT19
Rabbit
Antigen Affinity Chromatography
RUO
16669, 3880
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P08727, P19001
Krt19
Cytokeratin 19 Fusion Protein Ag1085
150 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers