missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CytokeRatin 18 Mouse anti-Human, CoraLite 555, Clone: 1G11C4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL555-66187-100UL
This item is not returnable.
View return policy
Description
CytokeRatin 18 Monoclonal antibody specifically detects CytokeRatin 18 in Human samples. It is validated for FC (Intra), IFSpecifications
| CytokeRatin 18 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| Mouse | |
| 100 μL | |
| 3875 | |
| Store at -20°C | |
| Liquid |
| Flow Cytometry, Immunofluorescence | |
| 1G11C4 | |
| CoraLite 555 | |
| BC006811 | |
| Protein A purification | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction