missing translation for 'onlineSavingsMsg'
Learn More

Cytohesin 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1710.33 NOK - 4836.10 NOK

Specifications

Antigen Cytohesin 3
Concentration 0.48 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16896394
View Documents
Proteintech
16687-1-AP-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16886394
View Documents
Proteintech
16687-1-AP-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GRP1 (general receptor for phosphoinositides-1) contains a Pleckstrin homology (PH) domain as well as a Sec7 domain. The PH domain has high binding affinity for phosphatidylinositol 3,4,5-trisphosphate Ptdlns(3,4,5)P3), while the Sec7 homology domain is responsible for catalyzing guanine nucleotide exchange of ADP-ribosylation factor (ARF) proteins. GRP1 co-localizes with ARF6 and catalyzes GTP/GDP exchange on ARF6. It is known to interact with Ptdlns(3,4,5)P3 localized to the plasma membrane in vitro and may also be a Ptdlns(3,4,5)P3 receptor. Additionally, GRP1 may regulate protein sorting and membrane trafficking through interaction with the guanosine triphosphate ARF, and may control cell adhesion through interaction with integrins.
Specifications

Specifications

Cytohesin 3
Western Blot
Unconjugated
Rabbit
Rat, Human, Mouse
O08967, O43739, P97696
116693, 19159, 9265
Cytohesin 3 Fusion Protein Ag10025
Primary
-20°C
CYTH3
0.48 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
ARNO3, CYTH3, cytohesin 3, GRP1, Protein ARNO3, PSCD3
Cyth3
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt