Learn More
Cytiva Cytodex 3 Gamma Microcarriers
Product Code. 15767207 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
30 g
300 g
3 kg
Unit Size:
300g
30g
3kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15767207 3 kg 3kg
15747207 30 g 30g
15757207 300 g 300g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15767207 Supplier Cytiva Supplier No. 17548803
 Restricted Item
Estimated Shipment: 24-05-2027
The microcarrier of choice for cells that are difficult to culture in vitro - particularly for cells with an epithelial-like morphology.
  • Cytodex 3 Gamma is designed by chemically coupling of a thin layer of denatured porcine collagen type 1 to the cross-linked dextran matrix.
  • As the collagen surface layer can be digested by a variety of proteolytic enzymes, harvest of such cells from the microcarriers can be performed with maintained high cell viability and membrane integrity.
  • This feature of Cytodex 3 Gamma is beneficial in the development of successful serial subcultivation protocols for scaling up culture volumes.
  • Applications include the production of large quantities of cells, viruses and recombinant cell products (e.g. interferon, enzymes, nucleic acids, hormones), studies on cell adhesion, differentiation and cell function, perfusion column culture systems, microscopy studies, harvesting mitotic cells, isolation of cells, membrane studies, storage and transport of cells, assays involving cell transfer and studies on uptage of labelled compounds
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Production of large quantities of cells, viruses and recombinant cell products, studies on cell adhesion, differentiation and cell function
Quantity 3 kg