missing translation for 'onlineSavingsMsg'
Learn More

Cytochalasin D, 98.2%, MP Biomedicals™

Product Code. 11492162 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 11492162

Brand: MP Biomedicals 0215077105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 22144-77-0
Molecular Formula C30H37NO6
Molecular Weight (g/mol) 507.627
InChI Key SDZRWUKZFQQKKV-DKLHZCHASA-N
Synonym cytochalasin d
PubChem CID 131668624
SMILES CC1CC=CC2C(C(=C)C(C3C2(C(C=CC(C1=O)(C)O)OC(=O)C)C(=O)NC3CC4=CC=CC=C4)C)O

Specifications

Color White
Assay Percent Range 0.982
Quantity 5 mg
Formula Weight 507.6
Physical Form Powder
Chemical Name or Material Cytochalasin D, For HPLC analysis
compliance-icons compliance-icons
Product Identifier
  • Cytochalasin D
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H300 + H310 + H330-Fatal if swallowed, in contact with skin or if inhaled.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P310-Immediately call a POISON CENTER/doctor/.
  • P320-Specific treatment is urgent (see on this label).
  • P361-Take off immediately all contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.