Learn More
Cytochalasin D, 98.2%, MP Biomedicals™
Product Code. 11482162 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. 11482162
Brand: MP Biomedicals 0215077101
Chemical Identifiers
CAS 22144-77-0
Molecular Formula C30H37NO6
Molecular Weight (g/mol) 507.627
InChI Key SDZRWUKZFQQKKV-DKLHZCHASA-N
Synonym cytochalasin d
PubChem CID 131668624
SMILES CC1CC=CC2C(C(=C)C(C3C2(C(C=CC(C1=O)(C)O)OC(=O)C)C(=O)NC3CC4=CC=CC=C4)C)O
Specifications
Color White
Assay Percent Range 0.982
Quantity 1 mg
Formula Weight 507.6
Physical Form Powder
Chemical Name or Material Cytochalasin D, For HPLC analysis
compliance-icons compliance-icons
Product Identifier
  • Cytochalasin D
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H300 + H310 + H330-Fatal if swallowed, in contact with skin or if inhaled.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P310-Immediately call a POISON CENTER/doctor/.
  • P320-Specific treatment is urgent (see on this label).
  • P361-Take off immediately all contaminated clothing.
  • P405-Store locked up.