Description
- Dissociation enzyme mixes (for research only) for liver are kits intended for the dissociation of liver tissues in single-cell sample preparation workflows
- The kits have been optimized for use with manual and semi-automated tissue disaggregation techniques
- Work effectively at 37°C
- Improved single cell viability and reduced cellular aggregation
- Ready-made solution for liver tissue - Optimized for the dissociation of the different segments that are operative in respective tissue
- Optimized for use with manual tissue disaggregation techniques and semi-automated techniques including the VIA Extractor™ tissue disaggregator
- Enzyme mix B (for liver) is recommended for use at 37°C as liver is very sensitive to cold
Specifications
Specifications
| Form | Lyophilized |
| Product Type | Cell Culture Dissociation Reagent |
| Sterility | Non-sterile |
| With Additives | None |
| Without Additives | None |