Learn More
Thermo Scientific Chemicals Cytidine-5'-triphosphate disodium salt, 97%
Product Code. 15434649
Click to view available options
Quantity:
500 mg
1 g
2 g
Unit Size:
1g
2g
500mg
Product Code. 15434649
Brand: Thermo Scientific Chemicals J62238.04
Request Bulk or Custom Format
CAS: 36051-68-0 | C9H14N3Na2O14P3 | 527.12 g/mol
Cytidine-5'-triphosphate disodium salt is a cation-permeable ligand gated ion channel (P2X purinergic receptor) agonist. It is also used as an organic intermediate, pharmaceutical intermediate, food additive and flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cytidine-5′-triphosphate disodium salt is a cation-permeable ligand gated ion channel (P2X purinergic receptor) agonist. It is also used as an organic intermediate, pharmaceutical intermediate, food additive and flavoring agent.
Solubility
Soluble in water. Insoluble in ethanol, chloroform and ether.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 36051-68-0
Molecular Formula C9H14N3Na2O14P3
Molecular Weight (g/mol) 527.12
MDL Number MFCD00078193
InChI Key NFQMDTRPCFJJND-WFIJOQBCSA-L
Synonym CTP
PubChem CID 131675548
IUPAC Name disodium;(2R,3R,4S,5R)-2-(4-amino-2-oxopyrimidin-1-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate
SMILES [Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]1O
Specifications
Color White
Flash Point 113°C (235°F)
Quantity 2 g
Solubility Information Soluble in water. Insoluble in ethanol,chloroform and ether.
Formula Weight 527.12
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Cytidine-5'-triphosphate disodium salt