missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytidine-5'-triphosphate disodium salt, 97%

Product Code. 15424649
Change view
Click to view available options
Quantity:
500 mg
1 g
2 g
Unit Size:
1g
2g
500mg
Product Code. Quantity unitSize
15424649 1 g 1g
15444649 500 mg 500mg
15434649 2 g 2g
3 options
This item is not returnable. View return policy

Product Code. 15424649

Brand: Thermo Scientific Chemicals J62238.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 36051-68-0 | C9H14N3Na2O14P3 | 527.12 g/mol

Cytidine-5'-triphosphate disodium salt is a cation-permeable ligand gated ion channel (P2X purinergic receptor) agonist. It is also used as an organic intermediate, pharmaceutical intermediate, food additive and flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cytidine-5′-triphosphate disodium salt is a cation-permeable ligand gated ion channel (P2X purinergic receptor) agonist. It is also used as an organic intermediate, pharmaceutical intermediate, food additive and flavoring agent.

Solubility
Soluble in water. Insoluble in ethanol, chloroform and ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36051-68-0
Molecular Formula C9H14N3Na2O14P3
Molecular Weight (g/mol) 527.12
MDL Number MFCD00078193
InChI Key NFQMDTRPCFJJND-WFIJOQBCSA-L
Synonym CTP
PubChem CID 131675548
IUPAC Name disodium;(2R,3R,4S,5R)-2-(4-amino-2-oxopyrimidin-1-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate
SMILES [Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]1O

Specifications

Color White
Flash Point 113°C (235°F)
Quantity 1 g
Solubility Information Soluble in water. Insoluble in ethanol,chloroform and ether.
Formula Weight 527.12
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Cytidine-5'-triphosphate disodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.