missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin C Antibody (Cyst13) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-05763JF585
This item is not returnable.
View return policy
Description
Cystatin C Monoclonal antibody specifically detects Cystatin C in Human samples. It is validated for ELISA,Immunoassay,Sandwich ELISA
Specifications
| Cystatin C | |
| Monoclonal | |
| Janelia Fluor 585 | |
| ARMD11, bA218C14.4 (cystatin C), cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, post-gamma-globulin | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Cystatin C purified from human urine. | |
| 0.2 mL | |
| Cancer, Epitope Tags, Signal Transduction | |
| 1471 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Immunoassay, Sandwich ELISA | |
| Cyst13 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction