Learn More
Cystatin A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15962-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16845374
  • 3828.53 NOK / 150µL
Estimated Shipment: 17-10-2023
Description
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins, and kininogens. This gene encodes a stefin that functions as a cysteine protease inhibitor, forming tight complexes with papain and the cathepsins B, H, and L. The protein is one of the precursor proteins of cornified cell envelope in keratinocytes and plays a role in epidermal development and maintece. Stefins have been proposed as prognostic and diagnostic tools for cancer.
Specifications
Specifications
Cystatin A
Polyclonal
Unconjugated
CSTA
CSTA, Cystatin A, cystatin A (stefin A), Cystatin AS, Stefin A, STF1, STFA
Rabbit
Antigen Affinity Chromatography
RUO
1475, 209294
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.43 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P01040, P56567
Csta
Cystatin A Fusion Protein Ag8733
150 μL
Primary
Human, Mouse