missing translation for 'onlineSavingsMsg'
Learn More

Cyproheptadine hydrochloride, MedChemExpress

Product Code. p-200099066
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
50mg
Product Code. Quantity unitSize
16488256 100 mg 100mg
16482149 1 g 1g
16474337 50 mg 50mg
16452116 500 mg 500mg
4 options
This item is not returnable. View return policy

Product Code. 16488256

Brand: Medchem Express HYB0366A

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Cyproheptadine hydrochloride is a potent and orally active 5-HT2A receptor antagonist, with antidepressant and antiserotonergic effects. Cyproheptadine hydrochloride has antiplatelet and thromboprotective activities. Cyproheptadine hydrochloride can be used for the research of thromboembolic disorders.

Cyproheptadine hydrochloride is a potent and orally active 5-HT2A receptor antagonist, with antidepressant and antiserotonergic effects. Cyproheptadine hydrochloride has antiplatelet and thromboprotective activities. Cyproheptadine hydrochloride can be used for the research of thromboembolic disorders.

Specifications

Chemical Name or Material Cyproheptadine hydrochloride
Physical Form Solid
CAS 969-33-5
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H22ClN
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 50 mg/mL (154.39 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.07 mM; Need ultrasonic)
SMILES CN1CC/C(CC1)=C2C3=CC=CC=C3C=CC4=CC=CC=C/24.Cl
Molecular Weight (g/mol) 323.86
Formula Weight 323.86
Percent Purity 99.98%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.