Learn More
CYP27A1 Mouse anti-Human, Unconjugated, Clone: 3G3F7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67045-1-IG-20UL
 View more versions of this product
Product Code. 16809445
  • 1710.33 NOK / 20µL
Description
The CYP27A1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CYP27A1. This antibody recognizes human, mouse, rat antigen. The CYP27A1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
CYP27A1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
3G3F7
Unconjugated
20 μL
1593
IgG1