Learn More
Abnova™ CYP19A1 (Human) Recombinant Protein
Product Code. p-7164160
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16126984
Brand: Abnova™ H00001588P01.10ug
Human CYP19A1 full-length ORF ( AAH22896, 1 a.a. - 181 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: MVLEMLNPIHYNITSIVPEAMPAATMPVLLLTGLFLLVWNYEGTSSIPGPGYCMGIGPLISHGRFLWMGIGSACNYYNRVYGEFMRVWISGEETLIISKSSSMFHIMKHNHYSSRFGSKLGLQCIGMHEKGIIFNNNPELWKTTRPFFMKALSGPGLVRMVTVCAESLKTHLLLFTPASVN
Specifications
Accession Number AAH22896
Gene ID (Entrez) 1588
Name cytochrome P450, family 19, subfamily A, polypeptide 1
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias ARO, ARO1, CPV1, CYAR, CYP19, MGC104309, P-450AROM
Gene Symbol CYP19A1
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer