Learn More
ACROBiosystems Cynomolgus Her2 / ErbB2 Protein, His Tag
Brand:  ACROBiosystems HE2-C52Hb-100ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16447619
  • 3799.50 NOK / 100µg
Estimated Shipment: 20-01-2023
Description
Human Epidermal growth factor Receptor 2 (HER2) is also called ERBB2, HER-2,HER-2 /neu, NEU, NGL,TKR1 and c-erb B2,and is a protein giving higher aggressiveness in breast cancers. It is a member of the ErbB protein family, more commonly known as the epidermal growth factor receptor family. HER2 is a cell membrane surface-bound receptor tyrosine kinase and is normally involved in the signal transduction pathways leading to cell growth and differentiation. HER2 is thought to be an orphan receptor, with none of the EGF family of ligands able to activate it. Approximately 30% of breast cancers have an amplification of the HER2 gene or overexpression of its protein product. Overexpression of this receptor in breast cancer is associated with increased disease recurrence and worse prognosis. HER2 appears to play roles in development, cancer, communication at the neuromuscular junction and regulation of cell growth and differentiation .
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 71.2 kDa. The protein migrates as 85-100 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications
Specifications
XP_005584091.2
71.2 kDa
100 µg
Thr 23 - Thr 652
ERBB2,CD340,HER-2/neu,HER2,MLN19,NEU,NGL,TKR1
Unconjugated
PBS, pH7.4
HEK293
Cynomolgus
Powder
>95% as determined by SDS-PAGE
Cynomolgus Her2, His Tag (HE2-C52Hb) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # XP_005584091.2).
-20°C
Her2
His Tag
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens