missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Cynomolgus FOLR2 Protein, His Tag

Brand:  ACROBiosystems FO2-C52H8-100ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 17106331

  • 3531.19 NOK / 100µg
Estimated Shipment: 04-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Folate receptor beta is also known as Folate receptor 2, FBP, FOLR2, BETA-HFR, FBP/PL-1, FR-BETA, FR-P3, and is a member of the folate receptor (FOLR) family. and mediate delivery of 5-methyltetrahydrofolate to the interior of cells. This protein has a 68% and 79% sequence homology with the FOLR1 and FOLR3 proteins, respectively. The FOLR2 protein was originally thought to exist only in placenta, but is also detected in spleen, bone marrow, and thymus. FOLR2 is predominantly expressed in placenta, cells of the neutrophilic lineage, and some CD34+ hematopoietic progenitor cells. It is upregulated on myeloid leukemias, head and neck squamous cell carcinomas, and several nonepithelial cancers. It is also upregulated on macrophages and monocytes at chronic inflammatory sites including rheumatoid arthritis synovium and glioblastoma. FOLR2 is a marker for macrophages generated in the presence of M-CSF, but not GM-CSF. Its expression correlates with increased folate uptake ability. Folate conjugates of therapeutic drugs are a potential immunotherapy tool to target tumor-associated macrophages.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa. The protein migrates as 33-43 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

26.5 kDa
Cynomolgus FOLR2, His Tag (FO2-C52H8) is expressed from human 293 cells (HEK293). It contains AA Thr 34 - His 245 (Accession # A0A2K5U027-1).
-20°C
FOLR2
Cynomolgus Monkey
50 mM Tris, 100 mM Glycine, pH7.5
HEK293
Bio-Markers and CD Antigens
100 µg
Thr 34 - His 245
FOLR2,BETA-HFR,FBP/PL-1,FR-BETA,FR-P3,FBP
Unconjugated
His Tag
Less than 1.0 EU / µg by the LAL method.
Powder
>90% as determined by SDS-PAGE
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers