Learn More
ACROBiosystems Cynomolgus Fc gamma RIIA / CD32a Protein, His Tag (SPR & BLI & HPLC verified)
3357.64 NOK - 33576.41 NOK
Specifications
Accession Number | NP_001270598.1 |
---|---|
Molecular Weight (g/mol) | 22.1 kDa |
Source | Cynomolgus CD32a, His Tag (SPR and BLI and HPLC verified) (CDA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Pro 208 (Accession # Q8SPW4-1). |
Immunogen | Gln 28 - Pro 208 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16457349
|
ACROBiosystems
CDA-C52H7-1MG |
1 mg |
33576.41 NOK
1mg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16447349
|
ACROBiosystems
CDA-C52H7-100UG |
100 μg |
4578.60 NOK
100µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17166171
|
ACROBiosystems
CDA-C52H7-50UG |
50 μg |
3357.64 NOK
50µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Receptors for the Fc region of IgG (Fc γ R) are members of the Ig superfamily that function in the activation or inhibition of immune responses. Three classes of human Fc γ Rs: RI (CD64), RII (CD32), and RIII (CD16), which generate multiple isoforms, are recognized. There are three genes for human Fcγ RII /CD32 (A, B, and C) and one for mouse Fcγ RII B (CD32B). CD32 is a low affinity receptor for IgG. The activating isoform, CD32A, is expressed on monocytes, neutrophils, platelets and dendritic cells. CD32A is expressed on many immune cell types (macrophage, neutrophil, eosinophils, platelets, dendritic cells and Langerhan cells), where inhibitory ITIMbearing receptors may also be coexpressed and coengaged by specific ligands. CD32A delivers an activating signal upon ligand binding, and results in the initiation of inflammatory responses including cytolysis, phagocytosis, degranulation and cytokine production. The responses can be modulated by signals from the coexpressed inhibitory receptors such as CD32B, and the strength of the signal is dependent on the ratio of expression of the activating and inhibitory receptors.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.1 kDa. The protein migrates as 27-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_001270598.1 | |
Cynomolgus CD32a, His Tag (SPR and BLI and HPLC verified) (CDA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Pro 208 (Accession # Q8SPW4-1). | |
-20°C | |
Fc gamma RIIA / CD32a | |
Cynomolgus Monkey | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Fc Receptors |
22.1 kDa | |
Gln 28 - Pro 208 | |
CD32a,FCGR2A,CD32,FCG2 ,FCGR2A1,IGFR2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by reduced SDS-PAGE |