Learn More
BRAND™ PTFE Magnetic Stirring Bars, Cylindrical Shape
Product Code. 15714037 Shop All Brand Products
Change view
Click to view available options
Length (Metric):
2 mm
3 mm
5 mm
6 mm
8 mm
10 mm
12 mm
13 mm
15 mm
20 mm
25 mm
30 mm
35 mm
40 mm
45 mm
50 mm
60 mm
70 mm
80 mm
108 mm
159 mm
Diameter (Metric):
1.5 mm
2 mm
3 mm
4.5 mm
6 mm
8 mm
10 mm
27 mm
Unit Size:
Each
Pack of 10
Product Code. Length (Metric) Diameter (Metric) unitSize
15714037 70 mm 10 mm Pack of 10
12030922 8 mm 1.5 mm Pack of 10
11844070 5 mm 2 mm Pack of 10
11732513 10 mm 3 mm Pack of 10
10727671 108 mm 27 mm Each
11854070 6 mm 3 mm Pack of 10
12020922 8 mm 3 mm Pack of 10
12010922 3 mm 3 mm Pack of 10
11854140 12 mm 4.5 mm Pack of 10
12040922 15 mm 4.5 mm Pack of 10
11804080 10 mm 6 mm Pack of 10
11758208 15 mm 6 mm Pack of 10
11752513 25 mm 6 mm Pack of 10
11833960 30 mm 6 mm Pack of 10
11762513 35 mm 6 mm Pack of 10
15704037 40 mm 8 mm Pack of 10
12050922 45 mm 8 mm Pack of 10
10094171 50 mm 8 mm Pack of 10
12060922 60 mm 10 mm Pack of 10
11864140 80 mm 10 mm Pack of 10
11894070 15 mm 1.5 mm Pack of 10
15221329 2 mm 2 mm Pack of 10
15601265 20 mm 6 mm Pack of 10
15241329 13 mm 3 mm Pack of 10
10019940 159 mm 27 mm Each
25 options
Product Code. 15714037
Brand: BRAND™ 137145
Only null left
Add to basket
Add to basket
Manufactured from PTFE. BRAND™ PTFE Magnetic Stirring Bars, Cylindrical Shape are suitable for universal applications. Rounded, smooth surface provides effective stirring action.
  • Manufactured from PTFE
  • Rounded, smooth surface provides effective stirring action.
  • Suitable for universal applications.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 10 mm
Length (Metric) 70 mm
Material PTFE
Quantity 10/Pk.
Shape Cylinder