Learn More
- Home
- Products
- Hotplates and Stirrers
- Stirring Bars and Rods
- BRAND™ PTFE Magnetic Stirring Bars, Cylindrical Shape
BRAND™ PTFE Magnetic Stirring Bars, Cylindrical Shape
Product Code. 12050922 Shop All Brand Products
Change view
Click to view available options
Length (Metric):
2 mm
3 mm
5 mm
6 mm
8 mm
10 mm
12 mm
13 mm
15 mm
20 mm
25 mm
30 mm
35 mm
40 mm
45 mm
50 mm
60 mm
70 mm
80 mm
108 mm
159 mm
Diameter (Metric):
1.5 mm
2 mm
3 mm
4.5 mm
6 mm
8 mm
10 mm
27 mm
Unit Size:
Each
Pack of 10
| Product Code. | Length (Metric) | Diameter (Metric) | unitSize |
|---|---|---|---|
| 12050922 | 45 mm | 8 mm | Pack of 10 |
| 12030922 | 8 mm | 1.5 mm | Pack of 10 |
| 11844070 | 5 mm | 2 mm | Pack of 10 |
| 11732513 | 10 mm | 3 mm | Pack of 10 |
| 10727671 | 108 mm | 27 mm | Each |
| 11854070 | 6 mm | 3 mm | Pack of 10 |
| 12020922 | 8 mm | 3 mm | Pack of 10 |
| 12010922 | 3 mm | 3 mm | Pack of 10 |
| 11854140 | 12 mm | 4.5 mm | Pack of 10 |
| 12040922 | 15 mm | 4.5 mm | Pack of 10 |
| 11804080 | 10 mm | 6 mm | Pack of 10 |
| 11758208 | 15 mm | 6 mm | Pack of 10 |
| 11752513 | 25 mm | 6 mm | Pack of 10 |
| 11833960 | 30 mm | 6 mm | Pack of 10 |
| 11762513 | 35 mm | 6 mm | Pack of 10 |
| 15704037 | 40 mm | 8 mm | Pack of 10 |
| 10094171 | 50 mm | 8 mm | Pack of 10 |
| 12060922 | 60 mm | 10 mm | Pack of 10 |
| 15714037 | 70 mm | 10 mm | Pack of 10 |
| 11864140 | 80 mm | 10 mm | Pack of 10 |
| 11894070 | 15 mm | 1.5 mm | Pack of 10 |
| 15221329 | 2 mm | 2 mm | Pack of 10 |
| 15601265 | 20 mm | 6 mm | Pack of 10 |
| 15241329 | 13 mm | 3 mm | Pack of 10 |
| 10019940 | 159 mm | 27 mm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Manufactured from PTFE
- Rounded, smooth surface provides effective stirring action.
- Suitable for universal applications.
Specifications
Specifications
| Diameter (Metric) | 8 mm |
| Length (Metric) | 45 mm |
| Material | PTFE |
| Quantity | 10/Pk. |
| Shape | Cylinder |