Learn More
BRAND™ PTFE Magnetic Stirring Bars, Cylindrical Shape
Shop All Brand Products
Click to view available options
Length (Metric):
2 mm
3 mm
5 mm
6 mm
8 mm
10 mm
12 mm
13 mm
15 mm
20 mm
25 mm
30 mm
35 mm
40 mm
45 mm
50 mm
60 mm
70 mm
80 mm
108 mm
159 mm
Diameter (Metric):
1.5 mm
2 mm
3 mm
4.5 mm
6 mm
8 mm
10 mm
27 mm
Unit Size:
Each
Pack of 10
Description
- Manufactured from PTFE
- Rounded, smooth surface provides effective stirring action.
- Suitable for universal applications.
Specifications
Specifications
| Diameter (Metric) | 6 mm |
| Length (Metric) | 15 mm |
| Material | PTFE |
| Quantity | 10/Pk. |
| Shape | Cylinder |
Product Content Correction
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido