Learn More
Lenz Laborglasinstrumente™ Cylindrical Reaction Vessels with Thermostatic Jacket
Product Code. 11512254
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
2000 mL
3 L
4 L
5 L
6 L
10 L
20 L
Includes:
Laboratory Flange 100
Laboratory Flange 150
Laboratory Flange 200
Laboratory Flange 60
Unit Size:
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Includes Unit Size
11512254 500 mL Laboratory Flange 100 Each
11522254 1000 mL Laboratory Flange 100 Each
11532254 2000 mL Laboratory Flange 100 Each
11542254 3 L Laboratory Flange 100 Each
11552254 4 L Laboratory Flange 100 Each
11512264 10 L Laboratory Flange 150 Each
11522264 20 L Laboratory Flange 150 Each
11562254 2000 mL Laboratory Flange 150 Each
11582254 4 L Laboratory Flange 150 Each
11572254 3 L Laboratory Flange 150 Each
11592254 5 L Laboratory Flange 150 Each
11502264 6 L Laboratory Flange 150 Each
11542264 10 L Laboratory Flange 200 Each
11582244 500 mL Laboratory Flange 60 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 11512254 Supplier Lenz Laborglasinstrumente™ Supplier No. 06011258
  • ONLINE EXCLUSIVE PRICE
    7550.00 NOK
    6309.84 NOK / Pack of 1
    Save 1240.16 NOK
    16% Off
Estimated Shipment: 07-01-2027
Excellent for containing reactants and suitable for vacuum purposes. Lenz Laborglasinstrumente™ Cylindrical Reaction Vessels with Thermostatic Jacket are manufactured from durable DURAN™ borosilicate glass.
  • With graduation and laboratory flange LF
  • With flat bottom and two connectors (with threads GL 14/18 or ball flanges KF 15/25)
  • Delivery with two plastic olives (GL 14/18) and screw caps
  • Corrosion-free and resistant to thermal shock
  • Not suitable for the use under positive pressure
Specifications
Description DURAN™ Tubing
Capacity (Metric) 500 mL
Material Borosilicate Glass, Duran Glass
Height (Metric) 180 mm
Includes Laboratory Flange 100
Product Type Reaction vessel
Shape Cylindrical
Bottom Shape Flat
Diameter (Metric) Outer 130 mm