Learn More
Lenz Laborglasinstrumente™ Cylindrical Reaction Vessel, Round Bottom
Product Code. p-8013648
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
2000 mL
3 L
4 L
5 L
6 L
10 L
20 L
Includes:
Laboratory Flange 100
Laboratory Flange 150
Laboratory Flange 200
Laboratory Flange 60
Unit Size:
Each
Product Code. Capacity (Metric) Includes unitSize
11542184 500 mL Laboratory Flange 100 Each
11552184 2000 mL Laboratory Flange 100 Each
10091602 1000 mL Laboratory Flange 100 Each
11562184 3 L Laboratory Flange 100 Each
11572184 4 L Laboratory Flange 100 Each
11532194 10 L Laboratory Flange 150 Each
11542194 20 L Laboratory Flange 150 Each
11582184 2000 mL Laboratory Flange 150 Each
11592184 3 L Laboratory Flange 150 Each
11502194 4 L Laboratory Flange 150 Each
11522194 6 L Laboratory Flange 150 Each
11512194 5 L Laboratory Flange 150 Each
11562194 10 L Laboratory Flange 200 Each
11572194 20 L Laboratory Flange 200 Each
11552194 6 L Laboratory Flange 200 Each
11522184 1000 mL Laboratory Flange 60 Each
16 options
Product Code. 11542184
Brand: Lenz Laborglasinstrumente™ 06001258
Only null left
Specifications
Description DURAN™ Tubing
Capacity (Metric) 500 mL
Material Borosilicate Glass, Duran Glass
Height (Metric) 160 mm
Includes Laboratory Flange 100
Product Type Reaction vessel
Shape Cylindrical
Bottom Shape Round
Diameter (Metric) Outer 105 mm