Learn More
Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels, with Glass Plug
Product Code. 10673891
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Cone Size:
14/23 NS
29/32 NS
Includes:
Pressure Equalizing Tube
w/o Pressure Equalizing Tube
Unit Size:
Pack of 2
Product Code. Capacity (Metric) Cone Size Includes unitSize
10673891 500 mL 29/32 NS Pressure Equalizing Tube Pack of 2
10294431 1000 mL 29/32 NS w/o Pressure Equalizing Tube Pack of 2
10403881 1000 mL 29/32 NS Pressure Equalizing Tube Pack of 2
10616331 100 mL 29/32 NS Pressure Equalizing Tube Pack of 2
10632551 100 mL 29/32 NS w/o Pressure Equalizing Tube Pack of 2
10303291 100 mL 14/23 NS w/o Pressure Equalizing Tube Pack of 2
10484631 250 mL 29/32 NS w/o Pressure Equalizing Tube Pack of 2
10313101 250 mL 29/32 NS Pressure Equalizing Tube Pack of 2
10210841 25 mL 14/23 NS w/o Pressure Equalizing Tube Pack of 2
10139320 50 mL 14/23 NS Pressure Equalizing Tube Pack of 2
10539241 50 mL 29/32 NS w/o Pressure Equalizing Tube Pack of 2
10644661 50 mL 14/23 NS w/o Pressure Equalizing Tube Pack of 2
10557321 50 mL 29/32 NS Pressure Equalizing Tube Pack of 2
13 options
Product Code. 10673891
Brand: Lenz Laborglasinstrumente™ 04355814
Only null left
Add to basket
Add to basket
For the safe, easy and accurate transfer of fluids in the laboratory. Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels, with Glass Plug are manufactured from durable DURAN™ borosilicate glass.
  • With NS socket and cone
  • With scale (approx. volume/mL)
  • With inscription label
  • Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric) 500 mL
Material Borosilicate Glass
Cone Size 29/32 NS
Graduated Yes
Includes Pressure Equalizing Tube
Product Type Dropping Funnel