Learn More
Thermo Scientific Chemicals Al-24 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 40 x 30 x 26;Vol (ml), 20
Product Code. 15483515
Change view
Click to view available options
Capacity (Metric):
6 mL
20 mL
40 mL
60 mL
110 mL
225 mL
600 mL
Diameter (Metric) Outer:
16mm
26mm
30mm
36mm
50mm
55mm
73mm
Diameter (Metric) Inner:
30mm
40mm
50mm
60mm
75mm
100mm
150mm
Height (Metric):
20mm
30mm
35mm
40mm
65mm
75mm
85mm
Unit Size:
Each
Product Code. Capacity (Metric) Diameter (Metric) Outer Diameter (Metric) Inner Height (Metric) unitSize
15483515 6 mL 16mm 30mm 20mm Each
15493515 20 mL 26mm 40mm 30mm Each
15403525 40 mL 30mm 50mm 35mm Each
15413525 60 mL 36mm 60mm 40mm Each
15423525 110 mL 50mm 75mm 75mm Each
15433525 225 mL 55mm 100mm 65mm Each
15443525 600 mL 73mm 150mm 85mm Each
7 options
Product Code. 15483515
Brand: Thermo Scientific Chemicals 032961.KF
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 30mm
Material Al-24
Quantity 1 Piece
Capacity (Metric) 6 mL
Diameter (Metric) Outer 16mm
Height (Metric) 20mm
Product Type Cylindrical Al-24 Crucibles
What is the maximum working temperature of Al-24 Tubes?
The maximum working temperature for the Al-24 Tube is 1950 degrees C.